1-Hexyl-3-methylimidazolium trifluoromethanesulphonate 95%
|
Warning
|
Synonyms:
1-Hexyl-3-methylimidazolium triflate
, 1-Hexyl-3-methylimidazolium Trifluoromethanesulphonate
H59210.14EA
363
EUR
H59210.14
1-Hexyl-3-methylimidazolium trifluoromethanesulphonate 95%
1-Hexyl-3-methylimidazolium trifluoromethanesulphonate
Formula:
C₁₁H₁₉F₃N₂O₃S MW: 316,34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03427621 CAS Number: 460345-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...