(R)-(-)-2-Oxothiazolidine-4-carboxylic acid

Supplier: Thermo Scientific

Warning

J62254.06EA 143 EUR
J62254.06 J62254.03
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid

Reagent for hepatic glutathione studies.

Formula: C₄H₅NO₃S
Melting Pt: 174 °C (dec.)
MDL Number: MFCD00066092
CAS Number: 19771-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR