6,7-Dimethoxy-4-quinazolone ≥97%

Supplier: Thermo Scientific

Synonyms: 6,7-Dimethoxy-4(3H)-quinazolinone

H58789.03EA 242 EUR
H58789.03 H58789.06
6,7-Dimethoxy-4-quinazolone ≥97%
6,7-Dimethoxy-4-quinazolone
Formula: C₁₀H₁₀N₂O₃
MW: 206,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01570147
CAS Number: 13794-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR