8-Bromoguanosine

Supplier: Thermo Scientific

Synonyms: 2-Amino-8-bromo-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one

J63940.06EA 80.3 EUR
J63940.06
8-Bromoguanosine
8-Bromoguanosine
Formula: C₁₀H₁₂BrN₅O₅
MW: 362,14 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037985
CAS Number: 4016-63-1
EINECS: 223-677-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR