2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2)

Supplier: Thermo Scientific

Synonyms: 2'-Deoxycytidine-5'-triphosphate disodium salt2′-Deoxycytidine-5′-Triphosphate Trisodium Salt

J65686.MBEA 45.3 EUR
J65686.MB J65686.MD
2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2)
2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2)
Formula: C₉H₁₄N₃Na₂O₁₃P₃
MW: 511,12
CAS Number: 102783-51-7
EINECS: 252-849-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR