Protein MW markers, BlueStep™ low range, pre-stained
K972-0.5MLEA 275 EUR
VWRCK972-0.5ML
Protein MW markers, BlueStep™ low range, pre-stained
BlueStep™ protein molecular weight marker, low range, pre-stained has 6 bands ranging from 19.0 to 120.0 kDa. Pre-stained bands are easily transferred to blotting membranes for use in Western blotting. There is sufficient material for 50 assays.
  • 6 bands ranging from 19.0 to 120.0 kDa
  • Monitor real-time running conditions during electrophoresis