Affinity coupling media, Thiopropyl Sepharose™ 6B
Supplier: Cytiva
17-0420-01EA 580 EUR
GEHE17-0420-01
Affinity coupling media, Thiopropyl Sepharose™ 6B
Thiopropyl Sepharose™ 6B medium is used for reversible immobilisation of molecules containing thiol groups under mild conditions to form mixed disulphides.
  • Reversible coupling of proteins and small thiolated ligands through thiol groups to Sepharose™ 6B via a small hydroxypropyl spacer arm
  • Contains reactive 2-thiopyridyl disulphide groups attached to Sepharose through a chemically stable either linkage
  • For immobilising low molecular weight substances
  • Separate thiol-containing proteins and peptides from non-thiol-containing proteins and peptides