Ion exchange chromatography media, CM Sephadex™
Supplier: Cytiva
Sephadex™
17-0210-01EA 366 EUR
17-0210-01 17-0220-01 17-0220-02 17-0210-02
Ion exchange chromatography media, CM Sephadex™
Protein Purification Media
CM Sephadex™ is a weak cation exchanger based on the well documented and well proven Sephadex™ base matrix.
  • Weak cation exchanger suitable batch techniques
  • High binding capacities