Precast gels, DALT Gel
Supplier: Cytiva
17-6002-36EA 823 EUR
17-6002-36
Precast gels, DALT Gel
Electrophoresis Gels Precast Gels
DALT Gel is a precast polyacrylamide gel bound to a plastic support film, which provides ease of handling and dimensional stability. It is used in conjunction with DALT buffer kit.