Select-a-dye mono-reactive dye packs
Supplier: Cytiva
PA15123EA 448.71 EUR
PA15123
Select-a-dye mono-reactive dye packs
Protein Linking and Labeling Systems
Select-a-dye mono-reactive dye packs enables a variety of labelling strategies using NHS ester, maleimide, or hydrazide to be investigated. Example protocols included with each dye pack.
Delivery information: Contains 0,5 mg of each labelling chemistry per pack.