PIG® LITE-DRI® Absorbent
Supplier: New Pig
PIG® Lite-Dri® LITE-DRI®
NEWPPLP201EA 26.9 EUR
NEWPPLP201
PIG® LITE-DRI® Absorbent
Spill Control Materials
Silica-free Lite-Dri Loose is half the weight of clay and 3 times more absorbent. Use less material and drastically reduce your labour and disposal costs.
  • Unlike clay, which is simply coated with liquid, fast-wicking recycled cellulose actually soaks up the spill the moment it touches
  • Three times more absorbent means you'll use less material on the spill, reducing your labour and disposal costs
  • Non-abrasive composition won't harm finished floors or damage expensive machinery
  • Light-weight absorbent is easier than clay to carry and use without injury
  • Absorbs and retains oils, coolants, solvents and water
  • Bag features a built-in handle for easy portability, handling and dispersal
  • Recycled cellulose is ideal when green products are desired or required
Can be incinerated after use to reduce waste or for fuels blending
Weight: 10 kg per bag.
Volume bag: 30 L