N,N'-Methylene-bisacrylamide, PlusOne™
Supplier: Cytiva
17-1304-02EA 166.22 EUR
GEHE17-1304-02
N,N'-Methylene-bisacrylamide, PlusOne™
PlusOne™ N,N'-Methylenebisacrylamide is a general cross-linker used with acrylamide for PAGE techniques. It is supplied as a dry powder.