TrackerTape
Supplier: Cytiva
RPN2050EA 108.78 EUR
RPN2050
TrackerTape
TrackerTape is an adhesive, waterproof tape that phosphoresces to give a written image on autoradiographs. It proves an effective way to permanently identify membranes, dried gels, and their corresponding autoradiographs.
  • Produces a permanent negative image in minutes
  • Avoids confusion by allowing samples to be matched to results
  • Ordinary marker pen is used to write on the tape
Delivery information: 10 sheets per pack