Glycerine 87%, colourless liquid, PlusOne™

Supplier: Cytiva

Synonyms: Glycerol1,2,3-Propanetriol

17-1325-01EA 77.43 EUR
17-1325-01
Glycerine 87%, colourless liquid, PlusOne™
Glycerine

PlusOne™ Glycerol is a stabilising medium for casting polyacrylamide gradients. In IEF, the pH gradient can be stabilised by introducing 10 - 15% glycerol. It also stabilises polyacrylamide gels for preservation.


  • Soluble in cold water and hot water

Formula: HOCH₂CH(OH)CH₂OH
MW: 92,09 g/mol
Density: 1,2 g/ml
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://ie.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR