Precision balances, Quintix®
Supplier: Sartorius
Quintix®
611-2698EA 2720 EUR
611-2698 611-2679 611-3866 611-2644 611-3125 611-4246 611-2682 611-2745 611-2684 611-2744 SARBQUINTIX6131CFR 611-2703 611-2640 611-2707 611-2686 611-2708 611-2643 611-2742 611-2680 611-2647 611-2683 611-2692 611-2646 611-2690 611-2697 611-2748 611-2689 611-2743 611-2706 611-2693 611-2638 611-2696 611-3127 611-4264 611-2639 611-2704 611-2685 611-2642 611-2691 611-4160 611-2705 611-2699 611-2710 611-2747 611-2695 611-3130 611-2641 611-3129 611-2711 611-2712 611-2687 611-2681 611-3867 611-2694 611-4273 611-2746 611-2649 611-3126 611-2688 611-2678 611-2709 611-3128 611-2749
Precision balances, Quintix®
Balances and Scales
A series of ergonomic balances designed for easy, intuitive operation, with a large touch screen, self-explanatory icons and plain text prompts. In addition to the standard applications, they include general weighing, density, percentage, check weighing, peak hold, counting and averaging. The Quintix® range also provides mixing, components, statistics and conversion funtionalities. Results can be transferred via a mini USB interface to a printer or a computer directly into Microsoft Excel® spreadsheet without additional software. Menu with a choice of 10 languages.
  • High contrast colour touch screen
  • Stabilisation times of just 1,5 to 2 seconds
  • isoCAL internal calibration and adjustment function
  • Draught shield on 1 mg balances with doors that glide open smoothly that can also be completely taken apart for easy cleaning
  • Supervisor lock to prevent any unintentional changes to settings
  • Optional, GLP-compliant YDP30 printer that is automatically detected by the Quintix® balance
Delivery information: Supplied with universal power adapter, in-use cover and application guide.