Portable sampler, PB 17
Supplier: WTW
WTWO000186EA 5900 EUR
WTWO000186
Portable sampler, PB 17
The portable sampler PB 17 with vacuum technology and its immensely compact build is also suitable for field use in difficult terrain. With its large utilizable sample capacity of 24×1 L segment bottles, it is particularly suitable for extensive monitoring tasks and load determination.
  • Energy saving vacuum technology
  • Very compact build
  • Large bottle capacity
The lower part of the device has a double-wall insulation and offers full sample protection and a passive cooling facility with dry ice or cold storage accumulators. The cover, which is equipped with a lock, offers protection against the elements and unauthorized access. Its easy-to-change rechargeable battery with plug connection prevents downtimes caused by battery charging and facilitates a high level of availability.
Delivery information: Portable microprocessor-controlled liquid sampler in plastic housing with vacuum dosing system and built-in rechargeable battery. Delivery includes 5 m suction tubing, screw fitting, tubing end piece and 24×1 L PE sample bottles. Not included: battery charger. The required accessories for PB 17 must be ordered separately.