3,5,6-Trichlorosalicylic acid
Supplier: LGC Standards PROMOCHEM
Synonyms:
3,5,6-Trichlorosalicylic Acid
PROOBP341EA
314
EUR
PROOBP341
3,5,6-Trichlorosalicylic acid
3,5,6-Trichlorosalicylic acid
Formula:
C₇H₃Cl₃O₃ MW: 241,46 g/mol |
MDL Number:
MFCD00075245 CAS Number: 40932-60-3 EINECS: 255-144-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...