(±)-Aspartic acid
Supplier: LGC Standards PROMOCHEM
Synonyms:
2-Aminobutanedioic acid,DL-Aspartic acid
MM0567.00
PROOMM0567.00EA
1380
EUR
PROOMM0567.00
(±)-Aspartic acid
(±)-Aspartic acid
Formula:
C₄H₇NO₄ MW: 133,10 g/mol |
MDL Number:
MFCD00063083 CAS Number: 617-45-8 EINECS: 210-513-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...