N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine

MAYBBTB01842.1EA 0 EUR
MAYBBTB01842.1
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine
Formula: C₁₂H₁₃F₃N₂O₂
MDL Number: MFCD00052908
CAS Number: 1692-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR