Cyclopentadienyltitanium trichloride 99%
|
Danger
|
ACRO215770025EA
85.4
EUR
ACRO215770025
ACRO215770100
Cyclopentadienyltitanium trichloride 99%
Cyclopentadienyltitanium trichloride
Formula:
C₅Cl₃Ti MW: 214,28 g/mol Melting Pt: 170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003724 CAS Number: 1270-98-0 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | Yellow to orange Crystalline powder or crystals |
Infrared spectrum | Conforms |
Elemental Analysis | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...