Cyclopentadienylzirconium trichloride ≥98%

Supplier: Alfa Aesar
Danger

20617.06EA 364 EUR
20617.06 20617.MD 20617.03
Cyclopentadienylzirconium trichloride ≥98%
Cyclopentadienylzirconium trichloride
Formula: C₅H₅Cl₃Zr
MW: 262,676 g/mol
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00015756
CAS Number: 34767-44-7
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR