Ethyl acetopyruvate 98%

Supplier: Thermo Scientific

Synonyms: Ethyl 2,4-dioxovalerate

ACRO214010050EA 59.3 EUR
ACRO214010050
Ethyl acetopyruvate 98%
Ethyl acetopyruvate
Formula: C₇H₁₀O₄
MW: 158,15
Boiling Pt: 101…103 °C (12 mmHg)
Melting Pt: 16…18 °C
Density: 1,12 g/cm³
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009124
CAS Number: 615-79-2
EINECS: 210-447-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4760 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR