Ethyl acetopyruvate

Supplier: Molekula

Synonyms: Ethyl 2,4-dioxovalerate

MOLE19827000-25GEA 0 EUR
MOLE19827000-25G
Ethyl acetopyruvate
Ethyl acetopyruvate
Formula: C₇H₁₀O₄
MW: 158,15
MDL Number: MFCD00009124
CAS Number: 615-79-2
EINECS: 210-447-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR