Carbosulphan

Supplier: Merck

32005-100MGEA 90.9 EUR
32005-100MG
Carbosulphan
Carbosulfan
Formula: C₂₀H₃₂N₂O₃S
MDL Number: MFCD00128058
CAS Number: 55285-14-8
EINECS: 259-565-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR