N-Phenylglycine

Supplier: Molekula

Synonyms: 2-Anilinoacetic acid

MOLEM36752536EA 0 EUR
MOLEM36752536
N-Phenylglycine
N-Phenylglycine
Formula: C₈H₉NO₂
MW: 151,17
MDL Number: MFCD00014009
CAS Number: 103-01-5
EINECS: 203-070-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR