Dysprosium(III) oxalate decahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Warning

14593.14EA 54.6 EUR
14593.14 14593.22
Dysprosium(III) oxalate decahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Dysprosium (III) oxalate decahydrate
Formula: C₆Dy₂O₁₂·10H₂O
MW: 769,21 (589,06 anhy)
Melting Pt: 40 °C (-H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00150482
CAS Number: 24670-07-3
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR