Iodoacetic anhydride 97%
Supplier: Thermo Scientific
|
Danger
|
ACRO307590010EA
203
EUR
ACRO307590010
ACRO307592500
Iodoacetic anhydride 97%
Iodoacetic anhydride
Formula:
C₄H₄I₂O₃ Melting Pt: 43…49 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001080 CAS Number: 54907-61-8 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 43 to 49 °C |
Titration by coupling | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...