Diclofenac sodium salt
Supplier: Molekula
Synonyms:
[o-(2,6-dichloroanilino) phenyl]acetic acid sodium salt
, Sodium [o-[(2,6-dichlorophenyl) amino] phenyl] acetate
, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt
, GP-45840
, 2-[(2,6-Dichlorophenyl)amino]-benzeneacetic acid monosodium salt
MOLE39643340-25GEA
170
EUR
MOLE39643340-25G
Diclofenac sodium salt
Diclofenac sodium salt
Formula:
C₁₄H₁₀Cl₂NNaO₂ MW: 318,1 g/mol |
MDL Number:
MFCD00082251 CAS Number: 15307-79-6 EINECS: 239-346-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...