4'-Hydroxydiclofenac
Supplier: Thermo Scientific
|
Danger
|
J64095.X0EA
544
EUR
J64095.X0
4'-Hydroxydiclofenac
4'-Hydroxydiclofenac
Formula:
C₁₄H₁₁Cl₂NO₃ MW: 312,15 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01671980 CAS Number: 64118-84-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...