4'-Hydroxydiclofenac, Supelco®

Supplier: Merck

32412-10MGEA 618 EUR
32412-10MG
4'-Hydroxydiclofenac, Supelco®
4'-Hydroxydiclofenac
Formula: C₁₄H₁₁Cl₂NO₃
MW: 312,15
MDL Number: MFCD01671980
CAS Number: 64118-84-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR