[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) bromide (12,9% Pd)

Supplier: Thermo Scientific
Warning

44980.03EA 115 EUR
44980.03 44980.06
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) bromide (12,9% Pd)
[1,1'-Bis(diphenylphosphino)ferrocene]palladium (II) bromide
Formula: C₃₄H₂₈Br₂FeP₂Pd
Storage Temperature: Ambient
MDL Number: MFCD09953448
CAS Number: 124268-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR