Carbamazepine
Supplier: Enzo Life Sciences
Synonyms:
5H-Dibenzo[b,f]azepine-5-carboxamide
ALEXALX550084G005EA
83.7
EUR
ALEXALX550084G005
Carbamazepine
Carbamazepine
Formula:
C₁₅H₁₂N₂O MW: 236,27 g/mol |
MDL Number:
MFCD00005073 CAS Number: 298-46-4 EINECS: 206-062-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...