Buffer, DIPSO, 0,2M solution, pH 8,0

J61388.AEEA 24 EUR
ALFAJ61388.AE ALFAJ61388.AK
Buffer, DIPSO, 0,2M solution, pH 8,0
CAS No.: 102783-62-0
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00070010
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR