Diosmin

Supplier: Roth Carl

ROTH6736.1EA 59.3 EUR
ROTH6736.1
Diosmin
Diosmin
Formula: C₂₈H₃₂O₁₅
MDL Number: MFCD00009772
CAS Number: 520-27-4
EINECS: 208-289-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR