Hesperidin

Supplier: Roth Carl

Synonyms: (S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one

ROTH9972.1EA 167 EUR
ROTH9972.1
Hesperidin
Hesperidin
Formula: C₂₈H₃₄O₁₅
MW: 610,56 g/mol
MDL Number: MFCD00075663
CAS Number: 520-26-3
EINECS: 208-288-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR