Bismuth titanium isopropoxide 5% (w/v) in 2-propanol

Supplier: Thermo Scientific
Danger


Packaged under argon in resealable ChemSeal™ bottles.

42840.AEEA 314 EUR
42840.AE 42840.AC
Bismuth titanium isopropoxide 5% (w/v) in 2-propanol
Bismuth titanium isopropoxide
Formula: C₂₁H₄₉BiO₇Ti
MW: 670,48
Storage Temperature: Ambient
MDL Number: MFCD01865878
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR