Lansoprazole

Supplier: LGC Standards PROMOCHEM

Synonyms: 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole

PROOMM0474.00EA 1420 EUR
PROOMM0474.00
Lansoprazole
Lansoprazole
Formula: C₁₆H₁₄F₃N₃O₂S
MW: 369,36 g/mol
MDL Number: MFCD00866873
CAS Number: 103577-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR