Tricyclazole, Supelco®

Supplier: Merck

Synonyms: 5-Methyl[1,2,4]triazolo[3,4-b][1,3]benzothiazole

M-Clarity™ quality level = MQ100

45808-10MGEA 103 EUR
45808-10MG
Tricyclazole, Supelco®
Tricyclazole
Formula: C₉H₇N₃S
CAS Number: 41814-78-2
EINECS: 255-559-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR