Ergocalciferol (vitamin D2, calciferol)

Supplier: Enzo Life Sciences

Synonyms: (S,Z)-3-((E)-2-((1R,3aS,7aR)-1-((2R,5R,E)-5,6-Dimethylhept-3-en-2-yl)-7a-methylhexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexanol

ALX460025G001
ALEXALX460025G001EA 66.2 EUR
ALEXALX460025G001
Ergocalciferol (vitamin D2, calciferol)
Ergocalciferol (Vitamin D2, Calciferol)
Formula: C₂₈H₄₄O
MW: 396,66 g/mol
MDL Number: MFCD00166988
CAS Number: 50-14-6
EINECS: 200-014-9
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR