Ergocalciferol (vitamin D2, calciferol)

Supplier: LGC Standards PROMOCHEM

PROOICRS0295EA 257 EUR
PROOICRS0295
Ergocalciferol (vitamin D2, calciferol)
Ergocalciferol (Vitamin D2, Calciferol)
Formula: C₂₈H₄₄O
MW: 396,66
MDL Number: MFCD00166988
CAS Number: 50-14-6
EINECS: 200-014-9
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR