1-Cyanomethyl-4,5-dicyano-2-(4-pyridyl)imidazole ≥97%

Supplier: Thermo Scientific

Danger

H54254.06EA 591 EUR
H54254.06 H54254.MD H54254.03
1-Cyanomethyl-4,5-dicyano-2-(4-pyridyl)imidazole ≥97%
1-Cyanomethyl-4,5-dicyano-2-(4-pyridyl)imidazole
Formula: C₁₂H₆N₆
Storage Temperature: Ambient
MDL Number: MFCD20265215
CAS Number: 1347815-36-4
UN: 3439
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR