1-Boc-4,5-dicyano-2-(3-pyridyl)imidazole ≥97%

Supplier: Thermo Scientific
Warning

H54303.06EA 879 EUR
H54303.06 H54303.MD H54303.03
1-Boc-4,5-dicyano-2-(3-pyridyl)imidazole ≥97%
1-Boc-4,5-dicyano-2-(3-pyridyl)imidazole
Formula: C₁₅H₁₃N₅O₂
Storage Temperature: Ambient
MDL Number: MFCD20265231
CAS Number: 1347815-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR