Theophyllin

Supplier: Molekula

Synonyms: 1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione

MOLEM47373244EA 0 EUR
MOLEM47373244
Theophyllin
Theophyllin
Formula: C₇H₈N₄O₂
MW: 180,16 g/mol
MDL Number: MFCD00079619
CAS Number: 58-55-9
EINECS: 200-385-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR