1,1,2-Trimethyl-1H-benz[e]indole 96%

Supplier: Thermo Scientific
Warning

ACRO421900250EA 63.5 EUR
ACRO421900250
1,1,2-Trimethyl-1H-benz[e]indole 96%
1,1,2-Trimethyl-1H-benz[e]indole
Formula: C₁₅H₁₅N
MW: 209,29
Melting Pt: 111…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00082627
CAS Number: 41532-84-7
EINECS: 255-429-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 117 °C
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR