1,1,2-Trimethyl-1H-benz[e]indole

Supplier: Molekula

MOLE69351493-5GEA 120 EUR
MOLE69351493-5G
1,1,2-Trimethyl-1H-benz[e]indole
1,1,2-Trimethyl-1H-benz[e]indole
Formula: C₁₅H₁₅N
MW: 209,29
MDL Number: MFCD00082627
CAS Number: 41532-84-7
EINECS: 255-429-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR