Ionomycin free acid

Supplier: Enzo Life Sciences

ALEXALX450006M001EA 152 EUR
ALEXALX450006M001 ALEXALX450006M005
Ionomycin free acid
Ionomycin
Formula: C₄₁H₇₂O₉
MDL Number: MFCD06798385
CAS Number: 56092-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR