4-Amino-1-boc-piperidine

Supplier: Apollo Scientific

Synonyms: tert-Butyl 4-amino-1-piperidinecarboxylateN-Boc-4-piperidineamine

APOSOR9845-100GEA 106 EUR
APOSOR9845-100G APOSOR9845
4-Amino-1-boc-piperidine
4-Amino-1-Boc-piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200,28
MDL Number: MFCD01076201
CAS Number: 87120-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR