Allantoin

Supplier: EDQM

EDQMA0349000EA 192 EUR
EDQMA0349000
Allantoin
Allantoin
Formula: C₄H₆N₄O₃
MW: 158,12
MDL Number: MFCD00005260
CAS Number: 97-59-6
EINECS: 202-592-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR