Anisomycin ≥97%

Danger

J62964.MFEA 216.7 EUR
J62964.MF
Anisomycin ≥97%
Anisomycin
Formula: C₁₄H₁₉NO₄
Melting Pt: 140…141 °C
Storage Temperature: Freezer
MDL Number: MFCD00077650
CAS Number: 22862-76-6
EINECS: 245-269-7
UN: 3462
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR