2'-Amino-4',5'-methylenedioxyacetophenone hydrochloride 98%
Supplier: Thermo Scientific
|
Warning
|
B21788.14EA
140
EUR
B21788.14
B21788.06
2'-Amino-4',5'-methylenedioxyacetophenone hydrochloride 98%
2'-Amino-4',5'-methylenedioxyacetophenone hydrochloride
Formula:
C₉H₁₀ClNO₃ MW: 215,64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00053022 CAS Number: 93983-01-8 EINECS: 301-288-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...